Assay Details
Gene Symbol: Gm26744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138627.1 | - | 1 - 2 | 148 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 36455 |
Gm36455 | -4.8kb | Sense | Chr. 16 - 21833291 - 21838656 on NCBI Build GRCm38 | |
Downstream | transmembrane protein 41a | Tmem41a |
2900010K02Rik, 5730578N08Rik |
88.6kb | Sense | Chr. 16 - 21934327 - 21947552 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
