Assay Details
Gene Symbol: Gm29536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039521.1 | - | 3 - 4 | 610 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
potassium voltage-gated channel, Isk-related subfamily, gene 4 |
Kcne4 |
AW487937, MiRP3 |
Anti-sense | Chr. 1 - 78816949 - 78820025 on NCBI Build GRCm38 | |
Downstream | secretogranin II | Scg2 |
Chgc, SgII |
485.8kb | Sense | Chr. 1 - 79434669 - 79440121 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
