Assay Details
Gene Symbol: Gm16973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046210.1 | 1 - 2 | 182 | 68 | ||
GenBank mRNA | AK045225.1 | - | 1 - 2 | 450 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | M-phase phosphoprotein 8 | Mphosph8 |
1500035L22Rik, 4930548G07Rik, AU040635 |
-1.7kb | Anti-sense | Chr. 14 - 56668240 - 56697430 on NCBI Build GRCm38 |
Downstream | paraspeckle protein 1 | Pspc1 |
5730470C09Rik, AI327109, AI449052, PSP1 |
21.2kb | Sense | Chr. 14 - 56722441 - 56778316 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
