Assay Details
Gene Symbol: Gm26651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049390.1 | - | 2 - 3 | 243 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth arrest and DNA-damage-inducible 45 gamma | Gadd45g |
AI327420, C86281, CR6, DDIT2, OIG37 |
-124.9kb | Sense | Chr. 13 - 51846675 - 51848475 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4921525O09 gene | 4921525O09Rik | 120.2kb | Sense | Chr. 13 - 52096741 - 52124160 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
