Assay Details
Gene Symbol: Gm32122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134720.1 | - | 2 - 3 | 216 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
glutamatic-oxaloacetic transaminase 2, mitochondrial |
Got2 |
AL022787, FABP-pm, Got-2, mAspAT |
-490.1kb | Sense | Chr. 8 - 95864137 - 95888365 on NCBI Build GRCm38 |
Downstream | predicted gene 15679 | Gm15679 | OTTMUSG00000025218 | 2.6mb | Anti-sense | Chr. 8 - 99007127 - 99032740 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
