Assay Details
Gene Symbol: A830021F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140437.1 | - | 477 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | aquaporin 4 | Aqp4 | WCH4 | -10.4kb | Anti-sense | Chr. 18 - 15389394 - 15410977 on NCBI Build GRCm38 |
Downstream | carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9 | Chst9 | 5430438D01Rik | 26.5kb | Anti-sense | Chr. 18 - 15451773 - 15760415 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
