Assay Details
Gene Symbol: Gm11754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK149260.1 | - | 1 - 2 | 246 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromobox 4 | Cbx4 |
MPc2, PC2 |
-20.8kb | Sense | Chr. 11 - 119077571 - 119086237 on NCBI Build GRCm38 |
Downstream |
TBC1 domain family, member 16 |
Tbc1d16 |
B930087K01, BC026530, Tcd1d16 |
31.6kb | Sense | Chr. 11 - 119143043 - 119228499 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
