Assay Details
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK161734.1 | - | 1 | 2256 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
methionine adenosyltransferase II, beta |
Mat2b |
1110064C04Rik, 2410018D16Rik, AI182287, AU022853, MAT-II, MATIIbeta, TGR |
-19.2kb | Sense | Chr. 11 - 40679314 - 40695203 on NCBI Build GRCm38 |
Downstream | NudC domain containing 2 | Nudcd2 | 16.5kb | Anti-sense | Chr. 11 - 40733661 - 40740046 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
