Assay Details
Gene Symbol: Gm34045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136051.1 | - | 5 - 6 | 409 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700064J06 gene | 1700064J06Rik | -32.1kb | Anti-sense | Chr. 10 - 119092905 - 119103107 on NCBI Build GRCm38 | |
Downstream | cullin associated and neddylation disassociated 1 | Cand1 |
2310038O07Rik, 6330512O03Rik, AI195005, AI846556, D10Ertd516e, Tp120a, mKIAA0829 |
62.9kb | Sense | Chr. 10 - 119198812 - 119240055 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
