Assay Details
Gene Symbol: Gm39321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136151.1 | - | 46 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
sortilin-related receptor, LDLR class A repeats-containing |
Sorl1 |
2900010L19Rik, AI596264, AW261561, LR11, SorLA, gp250, mSorLA |
-17.8kb | Anti-sense | Chr. 9 - 41968489 - 42124289 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930546K05 gene | 4930546K05Rik | 39.1kb | Anti-sense | Chr. 9 - 42182238 - 42209231 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
