Assay Details
Gene Symbol: Gm33251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144224.1 | - | 2 - 3 | 295 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AU022793 | AU022793 | -88.6kb | Sense | Chr. 15 - 39962649 - 39967515 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 9330182O14 gene | 9330182O14Rik | 5.4kb | Sense | Chr. 15 - 40134365 - 40150010 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
