Assay Details
Gene Symbol: Gm37359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144610.1 | - | 66 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
leucine, glutamate and lysine rich 1 |
Lekr1 |
EG546798, EG624866, Gm6534 |
-37.1kb | Anti-sense | Chr. 3 - 65659539 - 65832480 on NCBI Build GRCm38 |
Downstream | cyclin L1 | Ccnl1 |
2610030E23Rik, AU018493, Ccnl, ania-6a |
73.6kb | Sense | Chr. 3 - 65945912 - 65958971 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
