Assay Details
Gene Symbol: Gm37359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144610.1 | - | 375 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 8120 | Mir8120 |
Gm27637, mmu-mir-8120 |
-169.7kb | Anti-sense | Chr. 3 - 65659288 - 65659426 on NCBI Build GRCm38 |
Downstream | cyclin L1 | Ccnl1 |
2610030E23Rik, AU018493, Ccnl, ania-6a |
109.7kb | Sense | Chr. 3 - 65945912 - 65958971 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
