Assay Details
Gene Symbol: Gm38562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158824.1 | - | 77 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
gamma-aminobutyric acid (GABA) A receptor, subunit beta 1 |
Gabrb1 |
AW061132, B230208N19Rik, Gabrb-1 |
Anti-sense | Chr. 5 - 71658056 - 72149572 on NCBI Build GRCm38 | |
Downstream | COMM domain containing 8 | Commd8 |
C77770, D5Buc26e |
456.8kb | Sense | Chr. 5 - 72156576 - 72168183 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
