Assay Details
Gene Symbol: Gm21284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_078345.1 | 5 - 6 | 607 | 75 | ||
GenBank mRNA | AK137133.1 | - | 5 - 6 | 608 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | STAM binding protein | Stambp |
5330424L14Rik, 5730422L11Rik, AW107289, Amsh, mKIAA4198 |
-4.3kb | Anti-sense | Chr. 6 - 83543206 - 83575770 on NCBI Build GRCm38 |
Downstream |
C-type lectin domain family 4, member f |
Clec4f |
Clecsf13, D18063, KUCR_MOUSE, Kclr |
61.8kb | Anti-sense | Chr. 6 - 83644542 - 83656116 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
