Assay Details
Gene Symbol: 9430038I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029886.2 | NP_084162.1 | 2 - 3 | 281 | 76 | |
GenBank mRNA | AK020460.1 | - | 2 - 3 | 253 | 76 | |
AK051763.1 | - | 2 - 3 | 257 | 76 | ||
AK078159.1 | - | 2 - 3 | 261 | 76 | ||
BY148312.1 | - | 2 - 3 | 281 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
