Assay Details
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079869.1 | NP_001073338.1 | 2 - 3 | 411 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001079869.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
