Assay Details
Gene Symbol: Adora1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008533.3 | NP_001008533.1 | 1 - 2 | 1442 | 89 | |
NM_001039510.2 | NP_001034599.1 | 2 - 3 | 627 | 89 | ||
NM_001282945.1 | NP_001269874.1 | 2 - 3 | 661 | 89 | ||
GenBank mRNA | AJ555877.1 | - | 1 - 2 | 340 | 89 | |
AK038774.1 | - | 2 - 3 | 601 | 89 | ||
AK043954.1 | - | 1 - 2 | 1383 | 89 | ||
AK045729.1 | - | 1 - 2 | 982 | 89 | ||
AK134532.1 | - | 2 - 3 | 630 | 89 | ||
AK134553.1 | - | 1 - 2 | 847 | 89 | ||
AK134709.1 | - | 1 - 2 | 808 | 89 | ||
AU080986.1 | - | 2 - 3 | 568 | 89 | ||
BC026602.1 | - | 1 - 2 | 293 | 89 | ||
U05671.1 | - | 1 - 2 | 647 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
