Assay Details
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039700.2 | NP_001034789.2 | 11 - 12 | 2687 | 80 | |
NM_001286454.1 | NP_001273383.1 | 10 - 11 | 2657 | 80 | ||
NM_181544.2 | NP_853522.2 | 10 - 11 | 2657 | 80 | ||
GenBank mRNA | AB290926.1 | - | 10 - 11 | 2657 | 80 | |
AY164486.1 | - | 10 - 11 | 2657 | 80 | ||
DQ382344.1 | - | 10 - 11 | 2657 | 80 | ||
DQ382345.1 | - | 11 - 12 | 2687 | 80 | ||
DQ382346.1 | - | 11 - 12 | 2669 | 80 | ||
DQ382347.1 | - | 2 - 3 | 203 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
