Assay Details
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039700.2 | NP_001034789.2 | 30 - 31 | 6142 | 78 | |
NM_001286454.1 | NP_001273383.1 | 29 - 30 | 6112 | 78 | ||
NM_181544.2 | NP_853522.2 | 29 - 30 | 6112 | 78 | ||
GenBank mRNA | AB290926.1 | - | 29 - 30 | 6112 | 78 | |
AY164486.1 | - | 29 - 30 | 6112 | 78 | ||
DQ382344.1 | - | 29 - 30 | 6112 | 78 | ||
DQ382345.1 | - | 30 - 31 | 6142 | 78 | ||
DQ382349.1 | - | 11 - 12 | 1932 | 78 | ||
DQ382350.1 | - | 1932 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
