Assay Details
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008077.4 | NP_032103.2 | 5 - 6 | 776 | 104 | |
GenBank mRNA | AF326547.1 | - | 5 - 6 | 726 | 104 | |
AF326548.1 | - | 5 - 6 | 726 | 104 | ||
AF483492.1 | - | 4 - 5 | 543 | 104 | ||
AF483493.1 | - | 4 - 5 | 543 | 104 | ||
AK047521.1 | - | 5 - 6 | 912 | 104 | ||
AK136801.1 | - | 5 - 6 | 761 | 104 | ||
AK148497.1 | - | 5 - 6 | 776 | 104 | ||
AK158827.1 | - | 4 - 5 | 542 | 104 | ||
AK160410.1 | - | 4 - 5 | 686 | 104 | ||
AK160868.1 | - | 5 - 6 | 774 | 104 | ||
BC027059.1 | - | 5 - 6 | 727 | 5358787 | 104 | |
KC134210.1 | - | 4 - 5 | 543 | 104 | ||
KM102984.1 | - | 5 - 6 | 776 | 104 | ||
Y12257.1 | - | 4 - 5 | 543 | 104 | ||
Z49976.1 | - | 5 - 6 | 663 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
