Assay Details
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009025.2 | NP_033051.2 | 22 - 23 | 2387 | 73 | |
GenBank mRNA | AK053353.1 | - | 22 - 23 | 2383 | 73 | |
AK075768.1 | - | 8 - 9 | 1025 | 73 | ||
AK141511.1 | - | 22 - 23 | 2394 | 73 | ||
AK147896.1 | - | 22 - 23 | 2355 | 73 | ||
BC057300.1 | - | 22 - 23 | 2378 | 6821254 | 73 | |
BC060195.1 | - | 22 - 23 | 2402 | 73 | ||
BC068297.1 | - | 22 - 23 | 2251 | 30102545 | 73 | |
U20238.1 | - | 22 - 23 | 2372 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
