Assay Details
Gene Symbol: Cdc42ep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027219.3 | NP_081495.1 | 2 - 3 | 804 | 64 | |
GenBank mRNA | AK007896.1 | - | 2 - 3 | 805 | 64 | |
BC016250.1 | - | 2 - 3 | 792 | 4910994 | 64 | |
BC083130.1 | - | 2 - 3 | 797 | 6441478 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
