Assay Details
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139139.2 | NP_631878.2 | 7 - 8 | 557 | 109 | |
GenBank mRNA | AK077385.1 | - | 7 - 8 | 517 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
