Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252568.2 | NP_001239497.1 | 4 - 5 | 737 | 102 | |
NM_001252570.1 | NP_001239499.1 | 3 - 4 | 358 | 102 | ||
NM_001252571.1 | NP_001239500.1 | 3 - 4 | 358 | 102 | ||
NM_001281829.1 | NP_001268758.1 | 4 - 5 | 384 | 102 | ||
NM_172267.4 | NP_758471.1 | 4 - 5 | 737 | 102 | ||
GenBank mRNA | AI182929.1 | - | 4 - 5 | 359 | 102 | |
AK005293.1 | - | 4 - 5 | 738 | 102 | ||
AK138607.1 | - | 4 - 5 | 412 | 102 | ||
BC039982.1 | - | 4 - 5 | 379 | 102 | ||
BC132271.1 | - | 4 - 5 | 348 | 40130548 | 102 | |
BC132273.1 | - | 4 - 5 | 348 | 40130550 | 102 | |
BC145356.1 | - | 4 - 5 | 348 | 9053914 | 102 | |
BQ829083.1 | - | 3 - 4 | 354 | 102 | ||
CX235839.1 | - | 2 - 3 | 204 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
