Assay Details
Gene Symbol: S1pr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010101.4 | NP_034231.1 | 1 - 2 | 329 | 95 | |
GenBank mRNA | AB028143.1 | - | 1 - 2 | 197 | 95 | |
AK028043.1 | - | 1 - 2 | 186 | 95 | ||
AK029852.1 | - | 1 - 2 | 185 | 95 | ||
AK030134.1 | - | 1 - 2 | 184 | 95 | ||
AK047268.1 | - | 1 - 2 | 177 | 95 | ||
AK078443.1 | - | 1 - 2 | 183 | 95 | ||
AK081919.1 | - | 1 - 2 | 174 | 95 | ||
AK083405.1 | - | 1 - 2 | 163 | 95 | ||
AK084944.1 | - | 1 - 2 | 184 | 95 | ||
AK085180.1 | - | 1 - 2 | 184 | 95 | ||
AK087076.1 | - | 1 - 2 | 184 | 95 | ||
AK087139.1 | - | 1 - 2 | 182 | 95 | ||
BC064007.1 | - | 1 - 2 | 169 | 30244496 | 95 | |
BC068176.1 | - | 1 - 2 | 169 | 30613259 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB028143.1, AK028043.1, AK029852.1, AK030134.1, AK047268.1, AK078443.1, AK081919.1, AK084944.1, AK085180.1, BC064007.1, BC068176.1, NM_010101.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
