Assay Details
Gene Symbol: Mir185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029571.1 | 1 | 30 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transport and golgi organization 2 | Tango2 |
D16H22S680E, T10 |
Sense | Chr. 16 - 18300825 - 18348098 on NCBI Build GRCm38 | |
Downstream | armadillo repeat gene deleted in velo-cardio-facial syndrome | Arvcf | 20.7kb | Anti-sense | Chr. 16 - 18348182 - 18407076 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
