Assay Details
Gene Symbol: Mir148b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029766.1 | 1 | 47 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
coatomer protein complex, subunit zeta 1 |
Copz1 |
5930435A22Rik, AA407760, D4Ertd360e |
Sense | Chr. 15 - 103272834 - 103299865 on NCBI Build GRCm38 | |
Downstream | G protein-coupled receptor 84 | Gpr84 | EX33 | 23.0kb | Anti-sense | Chr. 15 - 103308233 - 103311242 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
