Assay Details
Gene Symbol: Mir10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029784.1 | 1 | 57 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox B3 | Hoxb3 | Hox-2.7 | Sense | Chr. 11 - 96307079 - 96347930 on NCBI Build GRCm38 | |
Downstream | homeobox B4 | Hoxb4 | Hox-2.6 | 993.0 | Sense | Chr. 11 - 96318267 - 96321638 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
