Assay Details
Gene Symbol: Mir222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029807.1 | 1 | 37 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 221 | Mir221 |
Mirn221, mir-221, mmu-mir-221 |
-505.0 | Sense | Chr. X - 19146294 - 19146388 on NCBI Build GRCm38 |
Downstream | uncharacterized LOC100503338 | LOC100503338 | 5.6kb | Sense | Chr. X - 19152588 - 19237927 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
