Assay Details
Gene Symbol: Mir455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030477.1 | 1 | 44 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin family member 12 | Kif12 | -84.7kb | Anti-sense | Chr. 4 - 63165629 - 63172142 on NCBI Build GRCm38 | |
Downstream | orosomucoid 1 | Orm1 |
Agp-1, Agp-2, Orm-1 |
87.6kb | Sense | Chr. 4 - 63344566 - 63348162 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
