Assay Details
Gene Symbol: Mir1839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_035501.1 | 1 | 38 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2900076A07 gene | 2900076A07Rik | WI1-1616M8.2 | Sense | Chr. 7 - 81523550 - 81531498 on NCBI Build GRCm38 | |
Downstream | fibronectin type III and SPRY domain containing 2 | Fsd2 |
9830160G03Rik, Spryd1 |
4.4kb | Anti-sense | Chr. 7 - 81534354 - 81566981 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
