Assay Details
Gene Symbol: Mir3103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037290.1 | 1 | 28 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
solute carrier family 5 (sodium/glucose cotransporter), member 2 |
Slc5a2 | Sglt2 | -15.9kb | Anti-sense | Chr. 7 - 128265697 - 128272433 on NCBI Build GRCm38 |
Downstream | regulator of G-protein signalling 10 | Rgs10 | 2310010N19Rik | 85.2kb | Sense | Chr. 7 - 128373621 - 128419061 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
