Assay Details
Gene Symbol: 2410018L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015504.1 | 2 - 3 | 455 | 115 | ||
GenBank mRNA | AK134640.1 | - | 2 - 3 | 456 | 115 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 4419 | Gm4419 | -1.5mb | Anti-sense | Chr. 12 - 21417707 - 21421064 on NCBI Build GRCm38 | |
Downstream | predicted gene 9292 | Gm9292 | EG668662 | 119.1kb | Sense | Chr. 12 - 23111957 - 23189721 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068406 | BC063063.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068406 | NR_015504.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068406 | AK134640.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976470 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068406 | BC063063.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068406 | NR_015504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068406 | AK134640.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976470 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10399672 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394845 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394815 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399641 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399673 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394806 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399643 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394824 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394933 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394808 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399656 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394932 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399674 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394842 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399647 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399644 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394854 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399660 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399646 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394822 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394826 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10394807 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10399645 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430845_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431317_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394852 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10399640 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394934 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394931 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10399659 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394827 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394837 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10399655 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10394940 | - | - |
Mouse Genome 430 2.0 Array | 1431317_at | - | - |
Mouse Genome 430 2.0 Array | 1431317_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430845_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430845_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69732_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69732_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200010567.mm.1 | TC1200001509.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200010567.mm.1 | NR_015504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010567.mm.1 | NR_015504.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010557.mm.1 | TC1200001509.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200010557.mm.1 | NR_015504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010557.mm.1 | NR_015504.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010566.mm.1 | TC1200001509.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200010566.mm.1 | NR_015504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010566.mm.1 | NR_015504.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010558.mm.1 | TC1200001509.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200010558.mm.1 | NR_015504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010558.mm.1 | NR_015504.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010564.mm.1 | TC1200001509.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200010564.mm.1 | NR_015504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200010564.mm.1 | NR_015504.1 | Refseq |
Mouse Expression Set 430 | 1439113_at | - | - |
Mouse Expression Set 430 | 1439113_PM_at | - | - |
Mouse Expression Set 430 | 1430845_at | - | - |
Mouse Expression Set 430 | 1430845_PM_at | - | - |
Murine Genome U74v2 Set | 104641_f_at | - | - |
Murine Genome U74v2 Set | 104641_PM_f_at | - | - |
Murine Genome U74v2 Set | 139031_f_at | - | - |
Murine Genome U74v2 Set | 139031_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1430845_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1439113_at | - | - |
Mouse Genome 430A 2.0 Array | 1430845_at | - | - |
Mouse Genome 430A 2.0 Array | 1439113_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430845_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430845_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431317_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431317_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738005 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2738005 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2542320 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2738005 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738005 | - | - |
