Assay Details
Gene Symbol: Gm12130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040295.1 | 1 - 2 | 187 | 87 | ||
GenBank mRNA | AK145334.1 | - | 1 - 2 | 157 | 87 | |
CJ052615.1 | - | 1 - 2 | 189 | 87 | ||
DV652764.1 | - | 1 - 2 | 131 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 12128 | Gm12128 | OTTMUSG00000005422 | -821.8kb | Anti-sense | Chr. 11 - 37658989 - 37673442 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930553C11 gene | 4930553C11Rik | 787.7kb | Sense | Chr. 11 - 39307703 - 39390225 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
