Assay Details
Gene Symbol: E130218I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040435.1 | 4 - 5 | 420 | 77 | ||
GenBank mRNA | CO429420.1 | - | 4 - 5 | 404 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | connector enhancer of kinase suppressor of Ras 1 | Cnksr1 | BC037758 | -6.5kb | Anti-sense | Chr. 4 - 134228041 - 134238416 on NCBI Build GRCm38 |
Downstream | glycine/arginine rich protein 1 | Grrp1 |
2810043G13Rik, Fam110d |
5.2kb | Anti-sense | Chr. 4 - 134251102 - 134266197 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
