Assay Details
Gene Symbol: Gm12505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040674.1 | 2 - 3 | 393 | 93 | ||
GenBank mRNA | BC049732.1 | - | 393 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RAD23b homolog (S. cerevisiae) | Rad23b |
0610007D13Rik, AV001138, HR23B, mHR23B, p58 |
-19.6kb | Anti-sense | Chr. 4 - 55350042 - 55392237 on NCBI Build GRCm38 |
Downstream | Kruppel-like factor 4 (gut) | Klf4 |
EZF, Gklf, Zie |
113.0kb | Sense | Chr. 4 - 55527137 - 55532475 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002777.rn.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
