Assay Details
Gene Symbol: 2610316D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045172.1 | 8 - 9 | 823 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DNA segment, Chr 3, ERATO Doi 751, expressed |
D3Ertd751e |
2810009O15Rik, 4930415G15Rik |
-3.5mb | Anti-sense | Chr. 3 - 41742606 - 41803292 on NCBI Build GRCm38 |
Downstream | protocadherin 10 | Pcdh10 |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
97.0kb | Anti-sense | Chr. 3 - 45378398 - 45434579 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2274383 | AK012035.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167858 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00631157 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892198 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892222 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892251 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892202 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00631212 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892100 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892236 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04892141 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00631189 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167858 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122748 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00631212 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167858 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04892222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02778477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101637 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02717761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22101493 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1458702_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457643_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1457643_x_at | - | - |
Mouse Genome 430 2.0 Array | 1458702_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457643_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1458702_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300014737.mm.1 | TC0300001920.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300014737.mm.1 | NR_045172.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014737.mm.1 | NR_045172.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014736.mm.1 | TC0300001920.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300014736.mm.1 | NR_045172.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300014736.mm.1 | NR_045172.1 | Refseq |
Mouse Expression Set 430 | 1457643_PM_x_at | - | - |
Mouse Expression Set 430 | 1458702_PM_at | - | - |
Mouse Expression Set 430 | 1457643_x_at | - | - |
Mouse Expression Set 430 | 1458702_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458702_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458702_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 784876 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102680128 | - | - |
