Assay Details
Gene Symbol: 1700042O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045178.1 | 1 - 2 | 72 | 100 | ||
GenBank mRNA | AK006690.1 | - | 1 - 2 | 73 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fidgetin-like 1 | Fignl1 | -59.1kb | Anti-sense | Chr. 11 - 11800288 - 11808995 on NCBI Build GRCm38 | |
Downstream | growth factor receptor bound protein 10 | Grb10 |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
59.3kb | Anti-sense | Chr. 11 - 11930499 - 12037422 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
