Assay Details
Gene Symbol: 1700064J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045348.1 | 1 - 2 | 29 | 59 | ||
GenBank mRNA | AK006882.1 | - | 1 - 2 | 30 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 33677 |
Gm33677 | -39.3kb | Anti-sense | Chr. 10 - 119028287 - 119053655 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 34045 |
Gm34045 | 41.4kb | Anti-sense | Chr. 10 - 119135154 - 119166385 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
