Assay Details
Gene Symbol: 4933405E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045506.1 | 1 - 2 | 452 | 92 | ||
GenBank mRNA | AK016664.1 | - | 1 - 2 | 455 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide |
P4ha2 |
AA407196, C76437, P4hl |
-77.0kb | Anti-sense | Chr. 11 - 54100924 - 54131668 on NCBI Build GRCm38 |
Downstream | colony stimulating factor 2 (granulocyte-macrophage) | Csf2 |
Csfgm, GMCSF, Gm-CSf, MGI-IGM |
34.0kb | Sense | Chr. 11 - 54247270 - 54249899 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
