Assay Details
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073167.1 | 1 - 2 | 787 | 75 | ||
GenBank mRNA | BC058699.1 | - | 1 - 2 | 762 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centromere protein P | Cenpp |
1700022C02Rik, 4921518G09Rik |
-199.0 | Anti-sense | Chr. 13 - 49464025 - 49652879 on NCBI Build GRCm38 |
Downstream | isoleucine-tRNA synthetase | Iars |
2510016L12Rik, AI327140, AU044614, E430001P04Rik, ILRS |
27.6kb | Sense | Chr. 13 - 49682130 - 49734267 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
