Assay Details
Gene Symbol: B020014A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045946.1 | 3 | 921 | 120 | ||
GenBank mRNA | AK139733.1 | - | 3 | 921 | 120 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 9230019H11 gene | 9230019H11Rik | Anti-sense | Chr. 10 - 3119812 - 3129546 on NCBI Build GRCm38 | ||
Downstream | histocompatibility 60c | H60c |
4632413I24Rik, Gm9491 |
129.4kb | Sense | Chr. 10 - 3256202 - 3268092 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
