Assay Details
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293715.1 | NP_001280644.1 | 8 - 9 | 2137 | 110 | |
NM_001293716.1 | NP_001280645.1 | 6 - 7 | 1155 | 110 | ||
NM_175356.3 | NP_780565.2 | 7 - 8 | 2092 | 110 | ||
GenBank mRNA | AK028974.1 | - | 7 - 8 | 2077 | 110 | |
AK030704.1 | - | 7 - 8 | 1852 | 110 | ||
AK048405.1 | - | 6 - 7 | 948 | 110 | ||
AK053614.1 | - | 7 - 8 | 2087 | 110 | ||
AK157918.1 | - | 7 - 8 | 1884 | 110 | ||
AK172074.1 | - | 7 - 8 | 2085 | 110 | ||
BC079846.1 | - | 7 - 8 | 2092 | 5707855 | 110 | |
BC113176.1 | - | 8 - 9 | 1992 | 40060773 | 110 | |
BC113781.1 | - | 6 - 7 | 1055 | 40060770 | 110 | |
BC138457.1 | - | 7 - 8 | 1995 | 8861477 | 110 | |
BC138458.1 | - | 7 - 8 | 1995 | 8861478 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
