Assay Details
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_075088.1 | 5 - 6 | 735 | 142 | ||
GenBank mRNA | AK160776.1 | - | 5 - 6 | 370 | 142 | |
BC028655.1 | - | 5 - 6 | 735 | 142 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peptidylprolyl isomerase domain and WD repeat containing 1 | Ppwd1 |
4632422M10Rik, A330090G21Rik |
-7.3kb | Anti-sense | Chr. 13 - 104204869 - 104228844 on NCBI Build GRCm38 |
Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 6 |
Adamts6 |
5031426K13, A930019D11Rik, ADAM-TS6, b2b1879.1Clo, b2b2029Clo, b2b2182Clo, b2b2187.1Clo, b2b2228Clo |
45.1kb | Sense | Chr. 13 - 104287347 - 104496694 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
