Assay Details
Gene Symbol: Dmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103477.1 | 6 - 7 | 571 | 82 | ||
GenBank mRNA | AK169570.1 | - | 6 - 7 | 571 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 | Ddx17 |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
-37.6kb | Sense | Chr. 15 - 79527696 - 79546764 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 227, member A |
Fam227a | 4933432B09Rik | 24.4kb | Sense | Chr. 15 - 79609576 - 79658970 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
