Assay Details
Gene Symbol: Gm12596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105031.1 | 2 - 3 | 169 | 80 | ||
GenBank mRNA | AK143878.1 | - | 2 - 3 | 170 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | UDP-glucose ceramide glucosyltransferase | Ugcg |
AU043821, C80537, Epcs21, GlcT-1, Ugcgl |
-37.8kb | Sense | Chr. 4 - 59189204 - 59222833 on NCBI Build GRCm38 |
Downstream | sushi domain containing 1 | Susd1 |
A530080P10, Gm12528, OTTMUSG00000007332 |
51.0kb | Anti-sense | Chr. 4 - 59314202 - 59439289 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
