Assay Details
Gene Symbol: Mir7k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105854.1 | 1 | 48 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pancreatic and duodenal homeobox 1 | Pdx1 |
IDX-1, IPF-1, Ipf1, Mody4, STF-1, pdx-1 |
-8.0kb | Anti-sense | Chr. 5 - 147270131 - 147275149 on NCBI Build GRCm38 |
Downstream | caudal type homeobox 2 | Cdx2 | Cdx-2 | 17.6kb | Sense | Chr. 5 - 147300900 - 147307249 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000160.rn.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
