Assay Details
Gene Symbol: Mir6540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105858.1 | 1 | 57 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | limbic system-associated membrane protein | Lsamp |
5430428I19, AW046674, B130007O04Rik, D930023J12Rik, Lam, Lamp |
-121.7kb | Anti-sense | Chr. 16 - 41532788 - 42181679 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4932412D23 gene | 4932412D23Rik |
ENSMUSG00000075070, Gm10809 |
422.2kb | Sense | Chr. 16 - 42725702 - 42875762 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
