Assay Details
Gene Symbol: Mir6955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105920.1 | 1 | 42 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26634 |
Gm26634 | -22.8kb | Anti-sense | Chr. 15 - 78862956 - 78869025 on NCBI Build GRCm38 | |
Downstream | SH3-domain binding protein 1 | Sh3bp1 | 3BP-1 | 7.9kb | Sense | Chr. 15 - 78899780 - 78912052 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
